N-[2-(Diethylamino)ethyl]-2,4-dimethyl-5-formyl-1H-pyrrole-3-carboxamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-{[2-(Diethylamino)ethyl]carbamoyl}-2,4-dimethyl-1H-pyrrole-2-carboxaldehyde, N-(2-(Diethylamino)ethyl)-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxamide, 3-{[2-(Diethylamino)ethyl]carbamoyl}-2,4-dimethyl-5-formyl-1H-pyrrole
A193863-100G
A193863-10G
A193863-1G
A193863-250MG
76873-156EA
401.03
USD
76873-156
76873-158
76873-160
76873-162
N-[2-(Diethylamino)ethyl]-2,4-dimethyl-5-formyl-1H-pyrrole-3-carboxamide 98%
N-[2-(Diethylamino)ethyl]-2,4-dimethyl-5-formyl-1H-pyrrole-3-carboxamide
Precaución: for Research Use Only
Formula:
C₁₄H₂₃N₃O₂ MW: 265.351 g/mol |
MDL Number:
MFCD12913910 CAS Number: 356068-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...