N2-(4-Fluorobenzyl)-5-nitropyridine-2,6-diamine 97%
Supplier: Ambeed
|
Warning
|
A193830-100G
A193830-100MG
A193830-1G
A193830-250MG
A193830-25G
A193830-5G
76873-144EA
92.3
USD
76873-144
76873-146
76873-148
76873-150
76873-152
76873-154
N2-(4-Fluorobenzyl)-5-nitropyridine-2,6-diamine 97%
N2-(4-Fluorobenzyl)-5-nitropyridine-2,6-diamine
Precaución: for Research Use Only
Formula:
C₁₂H₁₁FN₄O₂ MW: 262.23 g/mol |
MDL Number:
MFCD08282317 CAS Number: 33400-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...