5-Phenylthiazole 97%

Supplier: Ambeed
Warning

A193122-100MG A193122-1G A193122-250MG A193122-5G
76873-116EA 16.98 USD
76873-116 76873-118 76873-120 76873-122
5-Phenylthiazole 97%
5-Phenylthiazole

Precaución: for Research Use Only

Formula: C₉H₇NS
MW: 161.22 g/mol
MDL Number: MFCD12911591
CAS Number: 1826-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR