1,8-Dichloro-9,10-anthraquinone 97%

Supplier: Ambeed
Warning

Synonyms: 1,8-Dichloroanthraquinone, 1,8-Dichloroanthracene-9,10-dione

A192970-100G A192970-10G A192970-500G A192970-25G
76873-110EA 42.44 USD
76873-110 76873-112 76873-114 77084-998
1,8-Dichloro-9,10-anthraquinone 97%
1,8-Dichloro-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₆Cl₂O₂
MW: 277.11 g/mol
MDL Number: MFCD00001191
CAS Number: 82-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR