2-Amino-5-bromo-4-chlorobenzoic acid 97%

Supplier: Ambeed
Warning

A192012-1G A192012-250MG A192012-5G
76873-102EA 21 USD
76873-102 76873-104 76873-106
2-Amino-5-bromo-4-chlorobenzoic acid 97%
2-Amino-5-bromo-4-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
MDL Number: MFCD06409111
CAS Number: 50419-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR