1,1'-sulfonylbis(2-methyl-1H-imidazole) 97%

Supplier: Ambeed
Warning

A191388-1G A191388-250MG A191388-5G
76873-084EA 48.81 USD
76873-084 76873-086 76873-088
1,1'-sulfonylbis(2-methyl-1H-imidazole) 97%
1,1'-sulfonylbis(2-methyl-1H-imidazole)

Precaución: for Research Use Only

Formula: C₈H₁₀N₄O₂S
MW: 226.26 g/mol
MDL Number: MFCD18914518
CAS Number: 489471-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR