Methyl indole-3-carboxylate 98%

Supplier: Ambeed
Warning

A191313-100G A191313-10G A191313-25G A191313-500G A191313-5G
76873-074EA 30.77 USD
76873-074 76873-076 76873-078 76873-080 76873-082
Methyl indole-3-carboxylate 98%
Methyl indole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD00189407
CAS Number: 942-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR