4-(4-Bromopyridin-2-yl)piperazine, N1-BOC protected 95%

Supplier: Ambeed
Warning

A190922-1G A190922-5G
76873-046EA 20.16 USD
76873-046 76873-048 76873-050 76873-052
4-(4-Bromopyridin-2-yl)piperazine, N1-BOC protected 95%
4-(4-Bromopyridin-2-yl)piperazine, N1-BOC protected

Precaución: for Research Use Only

Formula: C₁₄H₂₀BrN₃O₂
MDL Number: MFCD13195778
CAS Number: 1197294-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR