1-(Difluoromethyl)-4-nitro-1H-pyrazole 98%

Supplier: Ambeed
Warning

A190353-5G A190353-25G
76873-034EA 45 USD
76873-034 77883-409
1-(Difluoromethyl)-4-nitro-1H-pyrazole 98%
1-(Difluoromethyl)-4-nitro-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₃F₂N₃O₂
MDL Number: MFCD04967412
CAS Number: 956477-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR