tert-Butyl (1-aminopropan-2-yl)carbamate 98%

Supplier: Ambeed
Warning

A190139-100MG A190139-10G A190139-1G A190139-250MG A190139-5G
76873-024EA 15 USD
76873-024 76873-026 76873-028 76873-030 76873-032
tert-Butyl (1-aminopropan-2-yl)carbamate 98%
tert-Butyl (1-aminopropan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₂
MDL Number: MFCD16660178
CAS Number: 149632-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR