(3-Bromo-2-methoxyphenyl)boronic acid 96%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-methoxyphenylboronic acid

A189738-1G A189738-250MG A189738-5G
76873-016EA 114.58 USD
76873-016 76873-018 76873-020
(3-Bromo-2-methoxyphenyl)boronic acid 96%
(3-Bromo-2-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₃
MW: 230.85 g/mol
MDL Number: MFCD18968533
CAS Number: 352525-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR