1-(4-Nitrophenyl)-2-piperidone 97%

Supplier: Ambeed
Warning

A189532-1G A189532-250MG A189532-25G A189532-5G
76873-014EA 17 USD
76873-014 77883-404 77883-405 77883-406
1-(4-Nitrophenyl)-2-piperidone 97%
1-(4-Nitrophenyl)-2-piperidone

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₃
MDL Number: MFCD04037154
CAS Number: 38560-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR