Di-tert-butyl piperazine-1,4-dicarboxylate 97%

Supplier: Ambeed
Warning

A188186-10G A188186-1G A188186-5G
76872-980EA 52 USD
76872-980 76872-982 76872-984
Di-tert-butyl piperazine-1,4-dicarboxylate 97%
Di-tert-butyl piperazine-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₆N₂O₄
MDL Number: MFCD01435774
CAS Number: 76535-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR