Boc-N-Me-D-Phe-OH 97%

Supplier: Ambeed
Warning

A187827-1G A187827-5G
76872-974EA 12 USD
76872-974 76872-976 76872-978
Boc-N-Me-D-Phe-OH 97%
Boc-N-Me-D-Phe-OH

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MDL Number: MFCD00151890
CAS Number: 85466-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR