2-Fluoro-1,3-dimethyl-5-nitrobenzene 97%

Supplier: Ambeed
Warning

A187625-10G A187625-1G A187625-25G A187625-5G
76872-966EA 124.13 USD
76872-966 76872-968 76872-970 76872-972
2-Fluoro-1,3-dimethyl-5-nitrobenzene 97%
2-Fluoro-1,3-dimethyl-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.153 g/mol
MDL Number: MFCD22055006
CAS Number: 1736-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR