N-((4-Methoxybenzyl)oxy)-2-nitrobenzenesulfonamide 97%

Supplier: Ambeed
Danger

A187310-10G A187310-1G A187310-250MG A187310-25G A187310-5G
76872-944EA 128.37 USD
76872-944 76872-946 76872-948 76872-950 76872-952
N-((4-Methoxybenzyl)oxy)-2-nitrobenzenesulfonamide 97%
N-((4-Methoxybenzyl)oxy)-2-nitrobenzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₂O₆S
MW: 338.34 g/mol
MDL Number: MFCD23099561
CAS Number: 1384122-86-4
UN: 2926
ADR: 4.1,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR