4-Amino-1-phenyl-1H-pyrazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A187279-100MG A187279-250MG
76872-940EA 45.62 USD
76872-940 76872-942
4-Amino-1-phenyl-1H-pyrazole-3-carboxylic acid 97%
4-Amino-1-phenyl-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉N₃O₂
MW: 203.20 g/mol
MDL Number: MFCD18809600
CAS Number: 64299-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR