1-Methyl-1H-benzimidazol-5-ol 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-benzo[d]imidazol-5-ol

A186938-100MG A186938-1G A186938-250MG A186938-5G A186938-10G A186938-25G
76872-930EA 16.98 USD
76872-930 76872-932 76872-934 76872-936 77611-810 77611-812
1-Methyl-1H-benzimidazol-5-ol 97%
1-Methyl-1H-benzimidazol-5-ol

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.162 g/mol
MDL Number: MFCD00456136
CAS Number: 50591-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR