4-Nitrobenzyl-(2S,4S)-2-(dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: (2S,4S)-4-Nitrobenzyl 2-(dimethylcarbamoyl)-4-mercaptopyrrolidine-1-carboxylate, (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylic Acid 4-Nitrobenzyl Ester, 4-Nitrobenzyl (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate

A186448-250MG A186448-5G A186448-100G A186448-1G
76872-922EA 5.31 USD
76872-922 76872-924 77611-800 77611-802
4-Nitrobenzyl-(2S,4S)-2-(dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate 98%
4-Nitrobenzyl-(2S,4S)-2-(dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₉N₃O₅S
MW: 353.39 g/mol
MDL Number: MFCD08063348
CAS Number: 96034-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR