3-Boronobenzenesulfonamide pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulphonamide

A186085-250MG A186085-1G
76872-894EA 50.93 USD
76872-894 77567-714
3-Boronobenzenesulfonamide pinacol ester 95%
3-Boronobenzenesulphonamide pinacol ester

Precaución: For research use only.

Formula: C₁₂H₁₈BNO₄S
MW: 283.1516 g/mol
MDL Number: MFCD07781217
CAS Number: 486422-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR