N-(Allyloxy)-2-nitrobenzenesulfonamide 95%

Supplier: Ambeed
Danger

A186024-100G A186024-10G A186024-1G A186024-250MG A186024-25G A186024-5G
76872-882EA 876.31 USD
76872-882 76872-884 76872-886 76872-888 76872-890 76872-892
N-(Allyloxy)-2-nitrobenzenesulfonamide 95%
N-(Allyloxy)-2-nitrobenzenesulfonamide

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₅S
MW: 258.25 g/mol
MDL Number: MFCD01413818
CAS Number: 359442-67-4
UN: 2926
ADR: 4.1,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR