8-Iodoquinazoline-2,4(1H,3H)-dione 98%

Supplier: Ambeed
Warning

A185660-100MG A185660-250MG
76872-876EA 36.08 USD
76872-876 76872-878
8-Iodoquinazoline-2,4(1H,3H)-dione 98%
8-Iodoquinazoline-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₈H₅IN₂O₂
MW: 288.04 g/mol
MDL Number: MFCD09954831
CAS Number: 959236-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR