O-(4-Methoxybenzyl)hydroxylamine 96%

Supplier: Ambeed
Danger

A184536-1G A184536-250MG
76872-864EA 80 USD
76872-864 77883-386
O-(4-Methoxybenzyl)hydroxylamine 96%
O-(4-Methoxybenzyl)hydroxylamine

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MDL Number: MFCD01722038
CAS Number: 21038-22-2
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR