2,6-Di(tert-butyl)-4-methylcyclohexan-1-ol 97%

Supplier: Ambeed
Warning

A184328-10G A184328-25G A184328-5G
76872-856EA 25 USD
76872-856 76872-858 76872-860 76872-862
2,6-Di(tert-butyl)-4-methylcyclohexan-1-ol 97%
2,6-Di(tert-butyl)-4-methylcyclohexan-1-ol

Precaución: for Research Use Only

Formula: C₁₅H₃₀O
MDL Number: MFCD00278493
CAS Number: 163119-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR