Ethyl 2-(3-piperidinylidene)acetate hydrochloride 98%

Supplier: Ambeed
Warning

A183754-100MG A183754-250MG
76872-846EA 54.11 USD
76872-846 76872-848
Ethyl 2-(3-piperidinylidene)acetate hydrochloride 98%
Ethyl 2-(3-piperidinylidene)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₆ClNO₂
MW: 205.68 g/mol
MDL Number: MFCD22421595
CAS Number: 2197130-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR