(S)-tert-Butyl (2-aminopropyl)carbamate hydrochloride 97%

Supplier: Ambeed
Warning

A183459-100MG A183459-250MG
76872-838EA 11 USD
76872-838 76872-840
(S)-tert-Butyl (2-aminopropyl)carbamate hydrochloride 97%
(S)-tert-Butyl (2-aminopropyl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₉ClN₂O₂
MDL Number: MFCD16295171
CAS Number: 1269493-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR