Methyl-3-chloro-1H-pyrrole-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-chloro-1H-pyrrole-2-carboxylate

A183312-100MG A183312-1G A183312-250MG A183312-5G A183312-10G
76872-824EA 6 USD
76872-824 76872-826 76872-828 76872-830 77883-381
Methyl-3-chloro-1H-pyrrole-2-carboxylate 95%
Methyl-3-chloro-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆ClNO₂
MDL Number: MFCD20661619
CAS Number: 226410-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR