2-Chloro-N-(2-(2-chlorobenzoyl)-4-nitrophenyl)acetamide 97%

Supplier: Ambeed
Danger

A183295-10G A183295-1G A183295-25G
76872-818EA 32 USD
76872-818 76872-820 76872-822
2-Chloro-N-(2-(2-chlorobenzoyl)-4-nitrophenyl)acetamide 97%
2-Chloro-N-(2-(2-chlorobenzoyl)-4-nitrophenyl)acetamide

Precaución: for Research Use Only

Formula: C₁₅H₁₀Cl₂N₂O₄
MDL Number: MFCD00799975
CAS Number: 180854-85-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR