Methyl 3-chloro-5-hydroxybenzoate 98%

Supplier: Ambeed
Warning

A182970-1G A182970-250MG
76872-808EA 250.38 USD
76872-808 76872-810
Methyl 3-chloro-5-hydroxybenzoate 98%
Methyl 3-chloro-5-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.592 g/mol
MDL Number: MFCD12407235
CAS Number: 98406-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR