Boc-Hyp-OEt 95%

Supplier: Ambeed
Warning

A182647-10G A182647-1G A182647-25G A182647-5G
76872-798EA 31.83 USD
76872-798 76872-800 76872-802 76872-804
Boc-Hyp-OEt 95%
Boc-Hyp-OEt

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₅
MW: 259.3 g/mol
MDL Number: MFCD09264314
CAS Number: 37813-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR