3-Fluoro-4-iodophenol 95%

Supplier: Ambeed
Warning

A181746-250MG A181746-1G A181746-5G A181746-10G
76872-778EA 24 USD
76872-778 77567-668 77611-768 77883-374
3-Fluoro-4-iodophenol 95%
3-Fluoro-4-iodophenol

Precaución: For research use only.

Formula: C₆H₄FIO
MDL Number: MFCD12407087
CAS Number: 122927-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR