Methyl-1,2,4-triazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A181362-100G A181362-10G A181362-25G A181362-500G A181362-5G
76872-762EA 10 USD
76872-762 76872-764 76872-766 76872-768 76872-770
Methyl-1,2,4-triazole-3-carboxylate 97%
Methyl 1,2,4-triazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₄H₅N₃O₂
MDL Number: MFCD00135989
CAS Number: 4928-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR