6-Cyanopyridine-2-boronic Acid Pinacol Ester 95%

Supplier: Ambeed
Warning

A181234-100MG A181234-250MG A181234-1G
76872-758EA 41 USD
76872-758 76872-760 77550-168
6-Cyanopyridine-2-boronic Acid Pinacol Ester 95%
6-Cyanopyridine-2-boronic Acid Pinacol Ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BN₂O₂
MDL Number: MFCD07367522
CAS Number: 952402-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR