3-Bromo-2-nitrophenol 95%

Supplier: Ambeed
Danger

A180708-100MG A180708-10G A180708-1G A180708-250MG A180708-5G
76872-742EA 9 USD
76872-742 76872-744 76872-746 76872-748 77883-372
3-Bromo-2-nitrophenol 95%
3-Bromo-2-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄BrNO₃
MDL Number: MFCD08445675
CAS Number: 76361-99-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR