5-Chloro-2,4-difluorobromobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-5-chloro-2,4-difluorobenzene, 5-Bromo-1-chloro-2,4-difluorobenzene

A180553-10G A180553-1G A180553-25G A180553-5G
76872-734EA 131.56 USD
76872-734 76872-736 76872-738 76872-740
5-Chloro-2,4-difluorobromobenzene 97%
5-Chloro-2,4-difluorobromobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClF₂
MW: 227.434 g/mol
MDL Number: MFCD04112557
CAS Number: 914636-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR