(4-(N-Ethylsulfamoyl)-2-methylphenyl)boronic acid 95%

Supplier: Ambeed
Warning

A180449-1G A180449-250MG
76872-732EA 207 USD
76872-732 77883-371
(4-(N-Ethylsulfamoyl)-2-methylphenyl)boronic acid 95%
(4-(N-Ethylsulfamoyl)-2-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₄BNO₄S
MW: 243.09 g/mol
MDL Number: MFCD13195667
CAS Number: 1217501-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR