Sodium (4-chloro-3-methylisoxazol-5-yl)((2-(2-(6-methylbenzo[d][1,3]dioxol-5-yl)acetyl)thiophen-3-yl)sulfonyl)amide 98%

Supplier: Ambeed
Warning

Synonyms: Sitaxsentan sodium

A180363-10MG A180363-1MG A180363-5MG
76872-722EA 84.88 USD
76872-722 76872-724 76872-726
Sodium (4-chloro-3-methylisoxazol-5-yl)((2-(2-(6-methylbenzo[d][1,3]dioxol-5-yl)acetyl)thiophen-3-yl)sulfonyl)amide 98%
Sodium (4-chloro-3-methylisoxazol-5-yl)((2-(2-(6-methylbenzo[d][1,3]dioxol-5-yl)acetyl)thiophen-3-yl)sulfonyl)amide

Precaución: for Research Use Only

Formula: C₁₈H₁₄ClN₂NaO₆S₂
MW: 476.89 g/mol
MDL Number: MFCD11040990
CAS Number: 210421-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR