N-Methyl-1-pyridin-2-ylmethanamine 98%

Supplier: Ambeed
Warning

Synonyms: 2-[(Methylamino)methyl]pyridine

A180067-100MG A180067-250MG A180067-5G
76872-716EA 10.61 USD
76872-716 76872-718 76872-720
N-Methyl-1-pyridin-2-ylmethanamine 98%
N-Methyl-1-pyridin-2-ylmethanamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD02089407
CAS Number: 21035-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR