cis-4-(Trifluoromethyl)cyclohexanecarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: cis-4-(Trifluoromethyl)cyclohexane-1-carboxylic acid

A179680-100MG A179680-1G
76872-702EA 23.34 USD
76872-702 77611-738
cis-4-(Trifluoromethyl)cyclohexanecarboxylic acid 95%
cis-4-(Trifluoromethyl)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₁F₃O₂
MW: 196.17 g/mol
MDL Number: MFCD09037361
CAS Number: 1202578-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR