AC-Thr(tBu)-OH 97%

Supplier: Ambeed
Warning

A179549-10G A179549-1G A179549-250MG
76872-696EA 170 USD
76872-696 76872-698 76872-700
AC-Thr(tBu)-OH 97%
AC-Thr(tBu)-OH

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MDL Number: MFCD00236771
CAS Number: 163277-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR