6-Bromo-5-chloro-1H-indole 98%
Supplier: Ambeed
|
Warning
|
A179153-1G
A179153-5G
A179153-250MG
A179153-100MG
76872-690EA
57
USD
76872-690
76872-692
77671-536
77883-360
6-Bromo-5-chloro-1H-indole 98%
6-Bromo-5-chloro-1H-indole
Precaución: for Research Use Only
Formula:
C₈H₅BrClN |
MDL Number:
MFCD11848570 CAS Number: 1191028-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...