(S)-4-(4-(5-(Aminomethyl)-2-oxooxazolidin-3-yl)phenyl)morpholin-3-one hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A179078-100G
A179078-10G
A179078-1G
A179078-250MG
A179078-25G
A179078-5G
76872-678EA
535
USD
76872-678
76872-680
76872-682
76872-684
76872-686
76872-688
(S)-4-(4-(5-(Aminomethyl)-2-oxooxazolidin-3-yl)phenyl)morpholin-3-one hydrochloride 97%
(S)-4-(4-(5-(Aminomethyl)-2-oxooxazolidin-3-yl)phenyl)morpholin-3-one hydrochloride
Precaución: for Research Use Only
Formula:
C₁₄H₁₈ClN₃O₄ MW: 327.76 g/mol |
MDL Number:
MFCD20133901 CAS Number: 898543-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...