3-chloro-4-hydroxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloropyridin-4-ol

A178975-1G A178975-5G
76872-674EA 23.34 USD
76872-674 76872-676
3-chloro-4-hydroxypyridine 98%
3-chloro-4-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄ClNO
MW: 129.543 g/mol
MDL Number: MFCD04114248
CAS Number: 89284-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR