(S)-tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A178966-100MG A178966-1G A178966-250MG
76872-668EA 7.43 USD
76872-668 76872-670 76872-672
(S)-tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate 98%
(S)-tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.27 g/mol
MDL Number: MFCD09832895
CAS Number: 790667-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR