5-Bromo-(2,3-methylenedioxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (6-Bromobenzo[d][1,3]dioxol-4-yl)boronic acid

A178562-1G A178562-250MG A178562-5G
76872-654EA 201.58 USD
76872-654 76872-656 76872-658
5-Bromo-(2,3-methylenedioxy)phenylboronic acid 98%
5-Bromo-(2,3-methylenedioxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BBrO₄
MW: 244.8400 g/mol
MDL Number: MFCD11855838
CAS Number: 1150114-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR