3-Bromo-5-fluoro-4-hydroxybenzaldehyde 95%

Supplier: Ambeed
Warning

A177980-100MG A177980-1G A177980-250MG A177980-5G
76872-630EA 21.22 USD
76872-630 76872-632 76872-634 76872-636
3-Bromo-5-fluoro-4-hydroxybenzaldehyde 95%
3-Bromo-5-fluoro-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
MW: 219.008 g/mol
MDL Number: MFCD21606586
CAS Number: 185345-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR