5-Bromo-2,4-difluorobenzoic acid 97%

Supplier: Ambeed
Warning

A177268-10G A177268-1G A177268-25G A177268-5G A177268-100G
76872-614EA 8.49 USD
76872-614 76872-616 76872-618 76872-620 76872-622 77671-518
5-Bromo-2,4-difluorobenzoic acid 97%
5-Bromo-2,4-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O₂
MDL Number: MFCD04115714
CAS Number: 28314-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR