Regorafenib 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-(3-(4-Chloro-3-(trifluoromethyl)phenyl)ureido)-3-fluorophenoxy)-N-methylpicolinamide, BAY-73-4506

A177150-1MG A177150-250MG A177150-5G A177150-25G
76872-610EA 6 USD
76872-610 76872-612 77611-716 77885-375
Regorafenib 98%
Regorafenib

Precaución: for Research Use Only

Formula: C₂₁H₁₅ClF₄N₄O₃
MDL Number: MFCD16038047
CAS Number: 755037-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR