4-Bromo-2-chloro-6-fluoroiodobenzene 97% stabilized

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1-chloro-3-fluoro-2-iodobenzene

Stabilization: Stabilized with Copper chip

A176890-1G A176890-250MG A176890-5G
76872-604EA 36.08 USD
76872-604 76872-606 77671-510
4-Bromo-2-chloro-6-fluoroiodobenzene 97% stabilized
4-Bromo-2-chloro-6-fluoroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFI
MW: 335.3400 g/mol
MDL Number: MFCD09878165
CAS Number: 83027-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR