3-Oxopyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester-2-ethyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl N-Boc-3-oxopyrrolidine-2-carboxylate, 1-tert-Butyl 2-ethyl 3-oxopyrrolidine-1,2-dicarboxylate

A176814-1G A176814-250MG A176814-25G A176814-5G
76872-598EA 148.53 USD
76872-598 76872-600 76872-602 77593-124
3-Oxopyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester-2-ethyl ester 95%
3-Oxopyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester-2-ethyl ester

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₅
MW: 257.29 g/mol
MDL Number: MFCD11052440
CAS Number: 170123-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR